missing translation for 'onlineSavingsMsg'
Learn More

Diacetamide-d7, TRC

Product Code. 30311161
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311161 5 mg 5mg
30321365 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30311161 Supplier Toronto Research Chemicals Supplier No. TRCA1345635MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetylacetamide-D7
CAS 33945-50-5
Molecular Formula C4 D7 N O2
Quantity 5 mg
Synonym Acetamide-N,2,2,2-d4, N-(acetyl-d3)- (9CI), Diacetamide-d7 (8CI), N-Acetylacetamide-D7, N-(Acetyl-d3)acetamide-N,2,2,2-d4
InChI Formula InChI=1S/C4H7NO2/c1-3(6)5-4(2)7/h1-2H3,(H,5,6,7)/i1D3,2D3/hD
SMILES [2H]N(C(=O)C([2H])([2H])[2H])C(=O)C([2H])([2H])[2H]
IUPAC Name N,2,2,2-tetradeuterio-N-(2,2,2-trideuterioacetyl)acetamide
Molecular Weight (g/mol) 108.147
Formula Weight 108.0916
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.