Learn More
Diacetin (Mixture), TRC
Specifications
Specifications
| Chemical Name or Material | Diacetin (Mixture) |
| CAS | 25395-31-7 |
| Molecular Formula | C7H12O5 |
| Quantity | 10 g |
| SMILES | CC(=O)OCC(CO)OC(=O)C |
| Molecular Weight (g/mol) | 176.17 |