missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diacetylclindamycin Phosphate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Diacetylclindamycin Phosphate
Specifications
Specifications
| Chemical Name or Material | Diacetylclindamycin Phosphate |
| Analyte or Component Names | Diacetylclindamycin Phosphate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C22 H38 Cl N2 O10 P S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Diacetylclindamycin Phosphate |
| InChI Formula | InChI=1S/C22H38ClN2O10PS/c1-7-8-14-9-15(25(5)10-14)21(28)24-16(11(2)23)17-18(32-12(3)26)19(33-13(4)27)20(22(34-17)37-6)35-36(29,30)31/h11,14-20,22H,7-10H2,1-6H3,(H,24,28)(H2,29,30,31)/t11-,14+,15-,16+,17+,18-,19-,20+,22+/m0/s1 |
| SMILES | CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@H](SC)[C@H](OP(=O)(O)O)[C@@H](OC(=O)C)[C@H]2OC(=O)C |
| IUPAC Name | [(2R,3S,4S,5R,6R)-3-acetyloxy-2-[(1S,2S)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-6-methylsulfanyl-5-phosphonooxyoxan-4-yl] acetate |
| Molecular Weight (g/mol) | 589.04 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction