Learn More
Diaion™ RCP160M(H), ion exchange resin, highly cross-linked, strongly acidic highly porous type, 1.5 meq/ml on PS-DVB, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046699.A1
Additional Details : Weight : 1.00000kg
Description
Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation. Used in solutions of high electrolyte content
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation
- Used in solutions of high electrolyte content
Specifications
250-710μm Beads | |
Highly cross-linked, strongly acidic highly porous type, 1.5meq/mL on PS-DVB | |
250 to 710μm |
Diaion™ RCP160M (H), ion exchange resin | |
Odorless | |
1 kg |
RUO – Research Use Only