Learn More
Diallylacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Diallylacetic Acid |
| CAS | 99-67-2 |
| Molecular Formula | C8 H12 O2 |
| Quantity | 25 mg |
| Synonym | 2-(2-Propen-1-yl)-4-pentenoic acid, 2-Allyl-4-pentenoic acid, 1,6-Heptadiene-4-carboxylic acid, 2-(2-Propenyl)-4-pentenoic acid, Di-2-propenylacetic acid, Diallylacetic acid, Valproic Acid Related Compound A |
| InChI Formula | InChI=1S/C8H12O2/c1-3-5-7(6-4-2)8(9)10/h3-4,7H,1-2,5-6H2,(H,9,10) |
| SMILES | OC(=O)C(CC=C)CC=C |
| IUPAC Name | 2-prop-2-enylpent-4-enoic acid |
| Molecular Weight (g/mol) | 140.18 |
| Formula Weight | 140.0837 |