Learn More
Diallylamine, 97%
Product Code. 15760789 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
Product Code. 15760789
Brand: Thermo Scientific Acros 112040010
Request Bulk or Custom Format
CAS: 124-02-7 | C6H11N | 97.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 124-02-7
Molecular Formula C6H11N
Molecular Weight (g/mol) 97.16
MDL Number MFCD00008642
InChI Key DYUWTXWIYMHBQS-UHFFFAOYSA-N
Synonym diallylamine, di-2-propenylamine, 2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, n-2-propenyl-2-propen-1-amine, unii-n18exb6v6p, n,n-diallylamine, ccris 4776, n-allylprop-2-en-1-amine
PubChem CID 31279
IUPAC Name N-prop-2-enylprop-2-en-1-amine
SMILES C=CCNCC=C
Specifications
Melting Point -88.0°C
Color Colorless to Yellow
Density 0.7890g/mL
Boiling Point 110.0°C to 112.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (H2C=CHCH2)2NH
Refractive Index 1.4395 to 1.4415
Quantity 1 L
Beilstein 04, 208
Merck Index 15, 2974
Specific Gravity 0.789
Solubility Information Solubility in water: 90g/L in water. Other solubilities: soluble in diethyl ether, ethanol
Formula Weight 97.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diallylamine, 97%