Learn More
Diamfenetide, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Diamfenetide |
| CAS | 36141-82-9 |
| Analyte or Component Names | Diamfenetide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Acetamide, N,N'-[oxybis(2,1-ethanediyloxy-4,1-phenylene)]bis-, Acetanilide, 4',4'''-[oxybis(ethyleneoxy)]bis- (6CI), Acemidophen, Coriban, Coryphamin, Diamfenetide, Diamphenethide, NSC 291838, WR 219124, Bis-beta-(4-acetamidophenoxy)ethyl ether |
| InChI Formula | InChI=1S/C20H24N2O5/c1-15(23)21-17-3-7-19(8-4-17)26-13-11-25-12-14-27-20-9-5-18(6-10-20)22-16(2)24/h3-10H,11-14H2,1-2H3,(H,21,23)(H,22,24) |
| Solution Type | Neat |
| SMILES | CC(=O)Nc1ccc(OCCOCCOc2ccc(NC(=O)C)cc2)cc1 |
| Show More |