Learn More
Surgical Design Glass Cutter with High Precision Diamond-Tip Inserts
Product Code. 10545234
Encompass_Preferred
Click to view available options
:
Glass Cutter
Product Code. 10545234
Brand: Surgical Design 33215
This item requires a price update prior to being ordered.
Request a Price for this item here.
Glass cutter designed for easy and effortless application. Bruce Diamond Corp. Diamond-inserts allows for easy and effortless application!
  • Diamond Glass Cutter Makes An Excellent Score Every Time
  • Leading industry diamond inserts by Bruce Diamond Corp
  • Made For The Experienced Glass Worker
  • Made From The Highest Quality Industrial Diamonds Available
Certifications
ASTM, ISO 9001, cGMP
Specifications
Material Brass
For Use With (Application) Breaking off single- or double-strength glass
Product Type Glass Cutter