missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DIAPH1 Recombinant Rabbit Monoclonal Antibody (6N10D0)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535137
This item is not returnable.
View return policy
Description
Immunogen sequence: MEPPGGSLGP GRGTRDKKKG RSPDELPSAG GDGGKSKKFT LKRLMADELE RFTSMRIKKE KEKPNSAHRN SSASYGDDPT AQSLQDVSDE QVLVLFEQML.
This gene is a homolog of the Drosophila diaphanous gene, and has been linked to autosomal dominant, fully penetrant, nonsyndromic sensorineural progressive low-frequency hearing loss. Actin polymerization involves proteins known to interact with diaphanous protein in Drosophila and mouse. It has therefore been speculated that this gene may have a role in the regulation of actin polymerization in hair cells of the inner ear. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| DIAPH1 | |
| Recombinant Monoclonal | |
| 0.95 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08808, O60610 | |
| DIAPH1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DIAPH1 (O60610). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6N10D0 | |
| Unconjugated | |
| DIAPH1 | |
| D18Wsu154e; DFNA1; DIA1; Diap1; Diaph1; diaphanous 1; diaphanous homolog 1; diaphanous homolog 1 (Drosophila); diaphanous related formin 1; diaphanous-1; diaphanous-related formin 1; Diaphanous-related formin-1; DRF1; FLJ25265; hDIA1; LFHL1; mDIA1; p140DIA; p140mDia; Protein diaphanous homolog 1; SCBMS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13367, 1729, 307483 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction