missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaphorase, MP BIOMEDICALS
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Application Notes:
- It has also been used in a study to assess the protein-protein interactions in assembly of lipoic acid on the 2-oxoacid dehydrogenases of aerobic metabolism. Lipoyl dehydrogenase has also been used in a study to investigate the redox regulation of tyrosine nitration and 3-nitrotyrosine reduction by antioxidants.
Usage Statement:
- Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Specifications
| Enzyme | Dehydrogenase |
| Quantity | 10 mg |
| Product Type | Lipoamide Dehydrogenase |
| Form | Freeze-dried (Lyophilized) |
Safety and Handling
Product Identifier
- Diaphorase
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Diaphorase
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction