Learn More
Diaveridine, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Diaveridine |
| CAS | 5355-16-8 |
| CAS Max % | ≤ 100% |
| Beilstein | 258464 |
| Color | Colorless |
| Melting Point | 235°C |
| Synonym | 5-(3,4-Dimethoxybenzyl)-2,4-pyrimidinediamine |
| Molecular Weight (g/mol) | 260.29 g/mol |
| Quantity | 100 mg |
| Grade | Coccidiostat |