missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaveridine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Diaveridine |
| CAS | 5355-16-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13 H16 N4 O2 |
| Quantity | 250 mg |
| Synonym | Pyrimidine, 2,4-diamino-5-veratryl- (6CI,7CI,8CI), 5-[(3,4-Dimethoxyphenyl)methyl]-2,4-pyrimidinediamine, 2,4-Diamino-5-(3,4-dimethoxybenzyl)pyrimidine, 2,4-Diamino-5-veratrylpyrimidine, BW 49-210, Diaveridin, Diaveridine, EGIS 5645, NSC 408735 |
| InChI Formula | InChI=1S/C13H16N4O2/c1-18-10-4-3-8(6-11(10)19-2)5-9-7-16-13(15)17-12(9)14/h3-4,6-7H,5H2,1-2H3,(H4,14,15,16,17) |
| SMILES | COc1ccc(Cc2cnc(N)nc2N)cc1OC |
| IUPAC Name | 5-[(3,4-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine |
| Molecular Weight (g/mol) | 260.29 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction