missing translation for 'onlineSavingsMsg'
Learn More

Diaveridine, TRC

Product Code. 30349195
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30349195

Brand: Toronto Research Chemicals TRCD41722510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diaveridine
CAS 5355-16-8
Purity Grade Notes HPLC
Molecular Formula C13 H16 N4 O2
Quantity 10 g
Synonym Pyrimidine, 2,4-diamino-5-veratryl- (6CI,7CI,8CI), 5-[(3,4-Dimethoxyphenyl)methyl]-2,4-pyrimidinediamine, 2,4-Diamino-5-(3,4-dimethoxybenzyl)pyrimidine, 2,4-Diamino-5-veratrylpyrimidine, BW 49-210, Diaveridin, Diaveridine, EGIS 5645, NSC 408735
InChI Formula InChI=1S/C13H16N4O2/c1-18-10-4-3-8(6-11(10)19-2)5-9-7-16-13(15)17-12(9)14/h3-4,6-7H,5H2,1-2H3,(H4,14,15,16,17)
SMILES COc1ccc(Cc2cnc(N)nc2N)cc1OC
IUPAC Name 5-[(3,4-dimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molecular Weight (g/mol) 260.29
Formula Weight 260.1273
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.