Learn More
Dibenzo-p-dioxin-d8, CDN
Product Code. 30314349
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314349
Brand: CDN CDND34730.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dibenzo-p-dioxin-d8
CAS 69699-83-8
Molecular Formula C12 2H8 O2
Quantity 0.01 g
Synonym Dibenzo[b,e][1,4 ]dioxin-1,2,3,4,6,7,8,9-d8 (9 CI), Dibenzo-p-dioxin D8
InChI Formula InChI=1 S/C12H8O2/c1-2-6-10-9(5-1)13-11-7-3-4-8-12(11)14-10/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D
SMILES [2 H]c1c([2 H])c([2 H])c2Oc3c([2 H])c([2 H])c([2 H])c([2 H])c3Oc2c1[2 H]
IUPAC Name 1,2,3,4,6,7,8,9-octadeuteriodibenzo-p-dioxin
Molecular Weight (g/mol) 192.24
Formula Weight 192.1026 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity