missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibromo-tyrosine Antibody (6G3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59350JF646
This item is not returnable.
View return policy
Description
Dibromo-tyrosine Monoclonal antibody specifically detects Dibromo-tyrosine in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Dibromo-tyrosine | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 5-dibromotyrosine;5-Dibromo-tyrosine;Dibromotyrosine;Dibromo-tyrosine;DiBrY | |
| Synthetic 3,5-Dibromotyrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 0.1 mL | |
| Cancer, Cellular Signaling | |
| Non-species specific | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 6G3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction