missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dicer Monoclonal Antibody (N167/7)
GREENER_CHOICE

Product Code. 16312114
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16312114 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16312114 Supplier Invitrogen™ Supplier No. MA527595

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-27595 was sufficient for detection of Dicer in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 215kDa. This antibody was formerly sold as clone S167-7.

Dicer is a double-stranded RNA (dsRNA) endoribonuclease playing a central role in short dsRNA-mediated post-transcriptional gene silencing. It cleaves naturally occurring long dsRNAs and short hairpin pre-microRNAs (miRNA) into fragments of twenty-one to twenty-three nucleotides with 3' overhang of two nucleotides, producing respectively short interfering RNAs (siRNA) and mature microRNAs. SiRNAs and miRNAs serve as guide to direct the RNA-induced silencing complex (RISC) to complementary RNAs to degrade them or prevent their translation. Gene silencing mediated by siRNAs, also called RNA interference, controls the elimination of transcripts from mobile and repetitive DNA elements of the genome but also the degradation of exogenous RNA of viral origin for instance. The miRNA pathway on the other side is a means to specifically regulate the expression of target genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dicer
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N167/7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene DICER1
Gene Accession No. Q8R418, Q9UPY3
Gene Alias 1110006F08Rik; D12Ertd7e; DCR; DCR1; DCR-1; Dicer; dicer 1 ribonuclease III; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease III; dicer 1, ribonuclease type III; Dicer1; Dicer1, Dcr-1 homolog; Dicer1e; double-strand-specific ribonuclease mDCR-1; endoribonuclease Dicer; Helicase MOI; helicase with RNAse motif; helicase-moi; HERNA; K12H4.8-LIKE; KIAA0928; mKIAA0928; MNG1; RMSE2
Gene Symbols DICER1
Host Species Mouse
Immunogen Fusion protein amino acids 1638-1899 of mouse Endoribonuclease Dicer.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192119, 23405, 299284
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.