missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diclazuril Polyclonal Antibody
Diclazuril Polyclonal Antibody
Brand: Invitrogen PA175109
This item is not returnable.
View return policy
Description
Diclazuril Polyclonal Antibody for ELISA
Specifications
| Diclazuril | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| Unconjugated | |
| C17H9Cl3N4O2 | |
| Diclazuril(4-N)-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction