missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DiD perchlorate
GREENER_CHOICE

Product Code. 18724719
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 18724719

Brand: Tocris Bioscience™ 5702/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipophilic fluorescent reagent; shown to stain HSCs

DiD perchlorate is a lipophilic fluorescent reagent; labels membranes and other hydrophobic structures. Shown to stain HSCs. Excitation/emission λ 644/665 nm.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[5-(1,3-Dihydro-3,3-dimethyl-1-octadecyl-2H-indol-2-ylidene)-1,3-pentadien-1-yl]-3,3-dimethyl-1-octadecyl-3H-indolium perchlorate
CAS 127274-91-3
Quantity 25 mg
Target Fluorescent Lipids Probes and Cell Membrane Stains
Molecular Formula C61H99ClN2O4
Formula Weight 959.9
Purity >95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.