missing translation for 'onlineSavingsMsg'
Learn More

Diethoxydimethylsilane, 97%

Product Code. 10530332 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10530332

Brand: Thermo Scientific Acros 174635000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-62-6 | C6H16O2Si | 148.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-62-6
MDL Number MFCD00009068
InChI Key YYLGKUPAFFKGRQ-UHFFFAOYSA-N
Synonym dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22
PubChem CID 62322
IUPAC Name diethoxy(dimethyl)silane
SMILES CCO[Si](C)(C)OCC

Specifications

Melting Point -70.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 114.0°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H16O2Si
Linear Formula (CH3)2Si(OC2H5)2
Refractive Index 1.3805 to 1.3825
Quantity 500 mL
Fieser 13,76
Specific Gravity 0.86
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 148.28
Formula Weight 148.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethoxydimethylsilane, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.