missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-Acetylmalonate, TRC

Product Code. 30385545
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385545 Supplier Toronto Research Chemicals Supplier No. TRCD4435131G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 2-Acetylmalonate
CAS 570-08-1
Molecular Formula C9H14O5
Quantity 1 g
Synonym Acetylmalonic Acid Diethyl Ester, C-Acetylmalonic Ester, Diethyl Acetylmalonate, Ethyl Acetomalonate, NSC 227297, NSC 4617
InChI Formula InChI=1S/C9H14O5/c1-4-13-8(11)7(6(3)10)9(12)14-5-2/h7H,4-5H2,1-3H3
SMILES CCOC(=O)C(C(=O)C)C(=O)OCC
Molecular Weight (g/mol) 202.2
Formula Weight 202.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.