Learn More
Diethyl (bromodifluoromethyl)phosphonate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 65094-22-6 |
| Molecular Formula | C5H10BrF2O3P |
| MDL Number | MFCD00069118 |
| Quantity | 1 g |
| SMILES | CCOP(=O)(OCC)C(F)(F)Br |
| Molecular Weight (g/mol) | 267.01 |
| Percent Purity | 97% |
| Chemical Name or Material | Diethyl (bromodifluoromethyl)phosphonate |