missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chlorophosphate, 95%, Thermo Scientific Chemicals

Catalog Number p-108895
22.80 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
814-49-3
C4H10ClO3P
172.55
MFCD00009075
LGTLXDJOAJDFLR-UHFFFAOYSA-N
diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate
13139
1-[chloro(ethoxy)phosphoryl]oxyethane
CCOP(=O)(OCC)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
814-49-3
C4H10ClO3P
172.55
MFCD00009075
LGTLXDJOAJDFLR-UHFFFAOYSA-N
diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate
13139
1-[chloro(ethoxy)phosphoryl]oxyethane
CCOP(=O)(OCC)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 81.0°C (6.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2POCl
Refractive Index 1.4160 to 1.4180
Quantity 25 g
Beilstein 01,332
Fieser 01,248; 03,98; 04,152; 05,217; 06,192; 08,171
Specific Gravity 1.19
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in chloroform
Formula Weight 172.55
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl chlorophosphate, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl chlorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'