Learn More
Diethyl Ethyl(1-methylbutyl)malonate, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl Ethyl(1-methylbutyl)malonate |
| CAS | 76-72-2 |
| Molecular Formula | C14H26O4 |
| Quantity | 5 g |
| Synonym | Ethyl(1-methylbutyl)-malonic Acid Diethyl Ester, Ethyl(alpha-methylbutyl)-malonic Acid Diethyl Ester, Ethyl(1-methylbutyl)-propanedioic Acid Diethyl Ester, NSC 10824 |
| InChI Formula | InChI=1S/C14H26O4/c1-6-10-11(5)14(7-2,12(15)17-8-3)13(16)18-9-4/h11H,6-10H2,1-5H3 |
| SMILES | CCCC(C)C(CC)(C(=O)OCC)C(=O)OCC |
| IUPAC Name | diethyl 2-ethyl-2-pentan-2-ylpropanedioate |
| Molecular Weight (g/mol) | 258.35 |
| Formula Weight | 258.1831 |