Learn More
Diethyl Ethylmalonate, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl Ethylmalonate |
| CAS | 133-13-1 |
| Molecular Formula | C9H16O4 |
| Quantity | 25 g |
| Synonym | Ethylpropanedioic Acid Diethyl Ester, 2-Ethylmalonic Acid Diethyl Ester, Diethyl 2-Ethylmalonate, Diethyl Ethylmalonate, Diethyl α-Ethylmalonate, Ethylpropanedioic Acid Diethyl Ester, NSC 8706 |
| InChI Formula | InChI=1S/C9H16O4/c1-4-7(8(10)12-5-2)9(11)13-6-3/h7H,4-6H2,1-3H3 |
| SMILES | CCOC(=O)C(CC)C(=O)OCC |
| Molecular Weight (g/mol) | 188.22 |
| Formula Weight | 188.1049 |