Learn More
Diethyl terephthalate, 98%, BLD Pharm™
Product Code. 31105918
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31105918 5 g
31103681 100 g
31096320 25 g
31100743 500 g
31099501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31105918 Supplier BLD Pharm Supplier No. BD1247715g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 636-09-9
Molecular Formula C12H14O4
MDL Number MFCD00039891
Quantity 5 g
SMILES O=C(OCC)C1=CC=C(C(OCC)=O)C=C1
Molecular Weight (g/mol) 222.24
Percent Purity 98%
Chemical Name or Material Diethyl terephthalate