Learn More
Diethylbenzene, mixture of isomers
Product Code. 15403197
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403197 100 mL 100mL
15413197 500 mL 500mL
15493187 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15403197 Supplier Thermo Scientific Alfa Aesar Supplier No. A19548.AE
Request Bulk or Custom Format
CAS: 25340-17-4 | C10H14
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25340-17-4
Molecular Formula C10H14
MDL Number MFCD00792900
Synonym Dowex J
Specifications
Density 0.87
Boiling Point 182°C to 184°C
Flash Point 55°C (131°F)
Refractive Index 1.495
Quantity 100 mL
UN Number UN2049
Beilstein 1903396
Formula Weight 134.22
Chemical Name or Material Diethylbenzene, mixture of isomers
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethylbenzene, mixture of isomers
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Diethylbenzenes