missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanomethylphosphonate, 96%, Thermo Scientific Chemicals

Catalog Number p-4258950
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2537-48-6
C6H12NO3P
177.14
MFCD00001893
KWMBADTWRIGGGG-UHFFFAOYSA-N
diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
75676
2-diethoxyphosphorylacetonitrile
CCOP(=O)(CC#N)OCC
This item is not returnable. View return policy
2537-48-6
C6H12NO3P
177.14
MFCD00001893
KWMBADTWRIGGGG-UHFFFAOYSA-N
diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
75676
2-diethoxyphosphorylacetonitrile
CCOP(=O)(CC#N)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 101.0°C to 102.0°C (0.4 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2CN
Refractive Index 1.4312 to 1.4332
Quantity 10 g
Fieser 01,250; 02,130; 03,96; 04,150; 13,15
Specific Gravity 1.09
Formula Weight 177.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethyl cyanomethylphosphonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl cyanomethylphosphonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'