missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphinic chloride, Thermo Scientific™

Product Code. 11362769
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362769 1 g 1g
15483747 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362769 Supplier Thermo Scientific Alfa Aesar Supplier No. H27681.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Dimethylphosphinic chloride was used in preparation of dimethyl-ethynylphosphine oxide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-37-4
Molecular Formula C4H10ClOP
Molecular Weight (g/mol) 140.55
MDL Number MFCD00013903
InChI Key KGENPKAWPRUNIG-UHFFFAOYSA-N
Synonym diethylphosphinic chloride, diethylphosphorylchloride, diethylphosphoryl chloride, phosphinic chloride,diethyl, diethylphosphinoyl chloride, acmc-20al83, diethylchlorophosphine oxide, phosphinic chloride, diethyl, 1-chloro ethyl phosphoryl ethane
PubChem CID 5195381
IUPAC Name 1-[chloro(ethyl)phosphoryl]ethane
SMILES CCP(Cl)(=O)CC

Specifications

Refractive Index 1.469
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 140.55
Chemical Name or Material Diethylphosphinic chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.