missing translation for 'onlineSavingsMsg'
Learn More

Diethylstilbestrol, MedChemExpress

Product Code. 16431136
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16431136 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431136 Supplier Medchem Express Supplier No. HY14598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Diethylstilbestrol (Stilbestrol), a synthetic nonsteroidal estrogen used in the treatment of menopausal and postmenopausal disorders.

Diethylstilbestrol (Stilbestrol), a synthetic nonsteroidal estrogen used in the treatment of menopausal and postmenopausal disorders.

Specifications

Chemical Name or Material Diethylstilbestrol
CAS 56-53-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H20O2
Quantity 200 mg
Synonym Stilbestrol
Solubility Information DMSO : 50 mg/mL (186.32 mM; Need ultrasonic)
SMILES OC1=CC=C(/C(CC)=C(CC)/C2=CC=C(O)C=C2)C=C1
Molecular Weight (g/mol) 268.35
Formula Weight 268.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.