missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Diethylstilbestrol Monoclonal Antibody (2E6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549709
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Diethylstilbestrol is a synthetic, nonsteroidal form of estrogen. A well-known teratogen and carcinogen, diethylstilbestrol inhibits the hypothalamic-pituitary-gonadal axis, thereby blocking the testicular synthesis of testosterone, lowering plasma testosterone, and inducing a chemical castration. Diethylstilbestrol is used in the treatment of menopausal and postmenopausal disorders. It was also used formerly as a growth promoter in animals. [National Center for Biotechnology Information. PubChem Compound Database; CID=448537].
Specifications
| Diethylstilbestrol | |
| Monoclonal | |
| 2.8 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 2E6 | |
| Unconjugated | |
| C18H20O2; Diethylstilbestrol; Distilbene; Stilbestrol; Stilboestrol | |
| Conjugate of DES hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction