missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Nutrient Agar 1.5%

Product Code. 11723453
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11723453

Brand: BD 269100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with blood or other enrichments for cultivation of fastidious microorganisms

Nutrient Agar 1.5% is a modification of Nutrient Agar. This medium is a slightly alkaline general-purpose medium. Since the medium contains 0.8% sodium chloride, it can be used as a base for enrichment with blood, ascitic fluid or other supplements for cultivating fastidious microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Form Powder
Product Type Agar
For Use With (Application) Used with Blood or other enrichments for cultivation of fastidious microorganisms
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.