Learn More
Faithful™ Differential Blood Cell Counter
Product Code. p-8619008
Encompass
Click to view available options
Dimensions (L x W x H):
250 x 80 x 50mm
320 x 80 x 50mm
Unit Size:
Each
Product Code. 11708731
Brand: Faithful™ DBC9
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manual incremental counter with a durable ABS plastic outer shell. Faithful™ Differential Blood Cell Counter is ideal for classifying and counting white blood cells.
  • ABS outer shell can resist collision.
  • Color charts of white blood cells are supplied.
  • Bell sound at 100% total record count.
  • Reset knob clears figures.
  • Quality certification: ISO 9001, ISO 13485
Specifications
No. of Keys 5 Keys
Dimensions (L x W x H) 320 x 80 x 50mm
Includes Totalizer