Learn More
Gilson™ Digital Dry Bath Block for NMR tubes
Product Code. 16358631 Shop All Gilson Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
16358631 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16358631 Supplier Gilson™ Supplier No. 36117270
Accessories for the Digital Dry Bath series.
Accessory for Digital Dry Bath Series.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Gilson Digital Dry Bath Series
Holds 30 x NMR tubes
Type Dry Bath Block