Learn More
TFA Dostmann Digital Indoor/Outdoor Thermometer
This compact digital indoor-outdoor thermometer also shows the maximum and minimum values. The application possibilities are manifold, for instance by its waterproof probe. The device is powered by a standard AAA battery (not included).
Brand: TFA Dostmann 30.1066.01
Additional Details : Weight : 0.40000kg
Description
This compact thermometer is ideal for simultaneous measurement of indoor and outdoor temperature. You can switch from the room temperature to the outdoor display with the push of a button. A constantly changing display (every 5 seconds) can also be set. The maximum and minimum values are saved and can be recalled. Two small magnets on the back adhere to metal surfaces. Alternatively, you can use the fold-out stand. When placed at the window, the sensor cable is simply led outside to measure the outdoor temperature. The application possibilities of thermometers with sensor cables are manifold. The waterproof probe also allows use in the car, refrigerator or aquarium. The device is powered by a standard AAA battery (not included).
- Simultaneous measurement of indoor and outdoor temperature
- Display manually switchable or automatically changing
- Max.-min. function
- With magnet and fold-out stand
- Waterproof cable sensor also for freezer, fridge or aquarium
Specifications
Plastic | |
Yes | |
LCD |