Learn More
Hach™ Digital Titrator Cartridge
Product Code. p-7115592
Click to view available options
Range:
100 to 4000 mg/L CaCO3
For Use With (Application):
Alkalinity Determination
Concentration:
1.6 N
Unit Size:
Each
Product Code. 12814883
Brand: Hach™ 1438901
For determinations using a compatible Digital Titrator. Hach™ Digital Titrator Cartridge is a versatile and economical option for providing fast, accurate results when and where you need them.
  • Digital Titrator methods use a high-precision dispensation device and cartridges of concentrated titrant to allow analyses in the lab, the plant or the field
  • Interchangeable cartridges allow you to change titrants in seconds, without cleaning or rinsing burets
  • Multiple parameter/range combinations are available
  • Concentrated titrants allow 40-100 typical titrations per cartridge
Specifications
Analytes Alkalinity / Acid-Base
Composition Sulfuric acid (H2SO4), 1.600N
Concentration 1.6 N
Format Cartridge
For Use With (Application) Alkalinity Determination
For Use With (Equipment) Digital Titrator
No. of Tests 40 to 100
Range 100 to 4000 mg/L CaCO3
Sample Size 100 mL
Storage Requirements Room temperature (10 to 25°C)
Test Method Phenolphthalein / Bromcresol green - methyl red
Type Titration Cartridge
Show More Show Less
compliance-icons
Product Identifier
  • Sulfuric Acid, 1,600N crtg
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid