Learn More
Diglycidyl Resorcinol Ether (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | Resorcinol diglycidyl ether |
| CAS | 101-90-6 |
| Molecular Formula | C12 H14 O4 |
| Quantity | 1 mL |
| Synonym | Oxirane, 2,2'-[1,3-phenylenebis(oxymethylene)]bis-, Benzene, m-bis(2,3-epoxypropoxy)- (6CI,7CI,8CI), 2,2'-[1,3-Phenylenebis(oxymethylene)]bis[oxirane], 1,3-Bis(2,3-epoxypropoxy)benzene, 1,3-Diglycidyloxybenzene, 1,3-Dihydroxybenzene diglycidyl ether, Diglycidyl resorcinol ether, NSC 76621, Resorcinol bis(2,3-epoxypropyl) ether, Resorcinol diglycidyl ether, Resorcinol glycidyl ether, m-Bis(2,3-epoxypropoxy)benzene, m-Bis(glycidyloxy)benzene |
| InChI Formula | InChI=1S/C12H14O4/c1-2-9(13-5-11-7-15-11)4-10(3-1)14-6-12-8-16-12/h1-4,11-12H,5-8H2 |
| SMILES | C(Oc1cccc(OCC2CO2)c1)C3CO3 |
| IUPAC Name | 2-[[3-(oxiran-2-ylmethoxy)phenoxy]methyl]oxirane |
| Molecular Weight (g/mol) | 222.24 |
| Formula Weight | 222.0892 |