missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin Antibody (611621) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7520JF669
This item is not returnable.
View return policy
Description
Digoxigenin Monoclonal antibody specifically detects Digoxigenin in Broad Species Reactivity samples. It is validated for Immunohistochemistry
Specifications
| Digoxigenin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| DIG | |
| KLH-coupled Digoxigenin | |
| RUO | |
| Broad Species Reactivity | |
| Liquid |
| Immunohistochemistry | |
| 611621 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction