missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Digoxin Monoclonal Antibody (26G10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514747
This item is not returnable.
View return policy
Description
MA5-14747 targets Digoxin in Western Blot, ELISA, IP and RIA applications and shows reactivity with labeled oligonucleotides and chemical samples. Product MA514747 is a smaller package size of MID0303 (formerly sold as a Seradyn product).
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
Specifications
| Digoxin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1 mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
| Mouse | |
| Ion-exchange chromatography | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
| 26G10 | |
| Unconjugated | |
| C41H64O14; DGX; Lanoxin | |
| Digoxin-BSA. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction