Learn More
Dihydroberberine, TRC
Specifications
Specifications
| Chemical Name or Material | Dihydroberberine |
| CAS | 483-15-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20 H19 N O4 |
| Quantity | 100 mg |
| Synonym | Dihydroberberine, 6H-Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine, 5,8-dihydro-9,10-dimethoxy-, Berberine, dihydro- (6CI,7CI), Berbine, 13,13a-didehydro-9,10-dimethoxy-2,3-(methylenedioxy)- (8CI), 7,8-Dihydroberberine, Dihydroberberine, Dihydroumbellatine, NSC 331264 |
| InChI Formula | InChI=1S/C20H19NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,7-9H,5-6,10-11H2,1-2H3 |
| SMILES | COc1ccc2C=C3N(CCc4cc5OCOc5cc34)Cc2c1OC |
| Molecular Weight (g/mol) | 337.369 |
| Formula Weight | 337.131 |
| Show More |