missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydrolipoamide Dehydrogenase/DLD Antibody (OTI6D5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70574PE
This item is not returnable.
View return policy
Description
Dihydrolipoamide Dehydrogenase/DLD Monoclonal specifically detects Dihydrolipoamide Dehydrogenase/DLD in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Dihydrolipoamide Dehydrogenase/DLD | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| DLD | |
| Full length recombinant protein of human Dihydrolipoamide Dehydrogenase/DLD (NP_000099) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI6D5 | |
| Flow Cytometry | |
| 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex), branched chain keto acid dehydrogenase complex, dihydrolipoamide dehydrogenase (E3 component of pyruvate dehydrogenase complex, dihydrolipoamide dehydrogenasemitochondrial, DLDH, E3, E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, EC 1.8.1, EC 1.8.1.4, GCSL, Glycine cleavage system L protein, glycine cleavage system protein L, lipoamide dehydrogenase, lipoamide reductase, lipoyl dehydrogenase | |
| Mouse | |
| Affinity Purified | |
| Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience | |
| 1738.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction