missing translation for 'onlineSavingsMsg'
Learn More

Diisodecyl Phthalate (Mixture of Branched chain Isomers) (>80%), TRC

Product Code. 30376453
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376453 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376453 Supplier Toronto Research Chemicals Supplier No. TRCD4553855ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-isodecyl ester
CAS 26761-40-0
Molecular Formula C28 H46 O4
Quantity 5 mL
Synonym 1,2-Benzenedicarboxylic acid, diisodecyl ester (9CI), Phthalic acid, diisodecyl ester (6CI,8CI), Isodecyl alcohol, phthalate (2:1) (8CI), Bis(isodecyl) phthalate, DIDP, Diisodecyl phthalate, Emkarate 1020, Hexaplas DIDP, Isodecyl phthalate, Jayflex DIDP, PX 120, Palatinol DIDP, Palatinol Z, Plasthall DIDP, Reomol DiDP, Sansocizer DIDP, Sicol 184, Vestinol DZ, (mixture of isomers)
InChI Formula InChI=1S/C28H46O4/c1-5-16-24(4)18-11-9-15-22-32-28(30)26-20-13-12-19-25(26)27(29)31-21-14-8-6-7-10-17-23(2)3/h12-13,19-20,23-24H,5-11,14-18,21-22H2,1-4H3
SMILES CCCC(C)CCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC(C)C
IUPAC Name 2-O-(6-methylnonyl) 1-O-(8-methylnonyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 446.66
Formula Weight 446.3396
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.