Learn More
Diisoheptylphthlate, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Diisoheptyl phthalate | 71888-89-6 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Diisoheptyl phthalate |
| Packaging | 2mL Amber Prescored Glass Ampule |
| CAS | 71888-89-6, 75-09-2 |
| CAS Min % | 0.1, 99.9 |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| CAS Max % | 0.1, 99.9 |
| Molecular Formula | C22H34O4 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| InChI Key | RKELNIPLHQEBJO-UHFFFAOYSA-N |
| Solvent or Matrix | Methylene Chloride |
| Show More |
Safety and Handling
Product Identifier
- Single Component Standard
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : dichloromethane, 1,2-benzenedicarboxylicacid
Product intended for laboratory use only