Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for Various Instruments
Click to view available options
Volume (Metric):
2.5 mL
5 mL
10 mL
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Diluter/Dispenser Syringe for various instruments
Specifications
Specifications
| Description | 60mm stroke, 6-32 UNC thread in plunger button |
| Length (Metric) | 60 mm |
| Volume (Metric) | 5 mL |
| For Use With (Equipment) | Various Instruments |