missing translation for 'onlineSavingsMsg'
Learn More

Dimepranol Acedoben, TRC

Product Code. 30004254
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004254 25 g 25g
17972295 2.5 g 2.5g
17994875 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004254 Supplier Toronto Research Chemicals Supplier No. TRCD46046525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61990-51-0
Molecular Formula C9 H9 N O3 . C5 H13 N O
Molecular Weight (g/mol) 282.34
Synonym Benzoic acid, 4-(acetylamino)-, compd. with 1-(dimethylamino)-2-propanol (1:1), 2-Propanol, 1-(dimethylamino)-, 4-(acetylamino)benzoate (salt) (9CI), Dimepranol acedoben, NSC 59001
IUPAC Name 4-acetamidobenzoic acid;1-(dimethylamino)propan-2-ol
SMILES CC(O)CN(C)C.CC(=O)Nc1ccc(cc1)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H9NO3.C5H13NO/c1-6(11)10-8-4-2-7(3-5-8)9(12)13;1-5(7)4-6(2)3/h2-5H,1H3,(H,10,11)(H,12,13);5,7H,4H2,1-3H3
Formula Weight 282.158
Chemical Name or Material DiP.PAcBA (N,N-Dimethylamino-2-propanol with p-acetamido benzoic acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.