Learn More
Dimethyl 2,6-Dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate (Nitrophenylpyridine Analogue of Nifedipine), Mikromol™
Description
Dimethyl 2,6-Dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate (Nitrophenylpyridine Analogue of Nifedipine)
Specifications
Specifications
| Chemical Name or Material | Dimethyl 2,6-Dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate |
| CAS | 67035-22-7 |
| Analyte or Component Names | Dimethyl 2,6-Dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H16 N2 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Nifedipine Imp. A (EP), Nifedipine Nitrophenylpyridine Analog, Dimethyl 2,6-Dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate, Nitrophenylpyridine Analogue of Nifedipine, Nifedipine Impurity A |
| InChI Formula | InChI=1S/C17H16N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8H,1-4H3 |
| SMILES | COC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2ccccc2[N+](=O)[O-] |
| IUPAC Name | dimethyl 2,6-dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate |
| Show More |