Learn More
Dimethyl 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Dimethyl 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Specifications
Specifications
| Chemical Name or Material | Dimethyl 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| CAS | 21881-77-6 |
| Analyte or Component Names | Dimethyl 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 N2 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dimethyl 2,6-Dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Nitrendipine Imp. B (EP), Nicardipine Hydrochloride Imp. C (EP), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3,5-dimethyl ester, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, dimethyl ester (9CI), 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(m-nitrophenyl)-, dimethyl ester (8CI), 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester, BAY-a 4339, Dimethyl 4-(m-nitrophenyl)-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, m-Nifedipine, Nicardipine Related Compound C, Nicardipine Hydrochloride Impurity C, Nitrendipine Impurity B, Nicardipine Impurity C |
| InChI Formula | InChI=1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-6-5-7-12(8-11)19(22)23/h5-8,15,18H,1-4H3 |
| SMILES | COC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC)C |
| IUPAC Name | dimethyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate |
| Show More |