missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl 4-(2-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| CAS | 43067-01-2 |
| Analyte or Component Names | dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 Cl N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dimethyl 4-(2-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. G (EP), Amlodipine Imp. G (EP), Amlodipine USP Related Compound C, Amlodipine USP RC C, Dimethyl 4-(2-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Related Compound C, Amlodipine Besilate Impurity G, Amlodipine Impurity G |
| InChI Formula | InChI=1S/C17H18ClNO4/c1-9-13(16(20)22-3)15(11-7-5-6-8-12(11)18)14(10(2)19-9)17(21)23-4/h5-8,15,19H,1-4H3 |
| Solution Type | Neat |
| SMILES | COC(=O)C1=C(C)NC(=C(C1c2ccccc2Cl)C(=O)OC)C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction