Learn More
Dimethyl 5-bromoisophthalate, 98%
Product Code. p-7033506
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11410150
Brand: Thermo Scientific Alfa Aesar H50629.03
Request Bulk or Custom Format
CAS: 51760-21-5 | C10H9BrO4 | 273.082 g/mol
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51760-21-5
Molecular Formula C10H9BrO4
Molecular Weight (g/mol) 273.082
MDL Number MFCD00078709
InChI Key QUJINGKSNJNXEB-UHFFFAOYSA-N
Synonym dimethyl 5-bromoisophthalate, 5-bromoisophthalic acid dimethyl ester, dimethyl-5-bromoisophthalate, 1,3-benzenedicarboxylic acid, 5-bromo-, dimethyl ester, 1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate, dimethyl5-bromoisophthalate, 5-bromo-isophthalic acid dimethyl ester, 1,3-benzenedicarboxylic acid, 5-bromo-, 1,3-dimethyl ester, methyl 5-bromo-3-methoxycarbonyl benzoate, 5-bromoisophthalic acid, dimethyl ester
PubChem CID 103954
IUPAC Name dimethyl 5-bromobenzene-1,3-dicarboxylate
SMILES COC(=O)C1=CC(=CC(=C1)Br)C(=O)OC
Specifications
Melting Point 88°C to 90°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 273.08
Percent Purity 98%
Chemical Name or Material Dimethyl 5-bromoisophthalate