Learn More
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals
Catalog Number p-117449
39.73 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
95-45-4
C4H8N2O2
116.12
MFCD00002117
JGUQDUKBUKFFRO-GGWOSOGESA-N
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
5356010
C\C(=N/O)\C(\C)=N\O
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
95-45-4
C4H8N2O2
116.12
MFCD00002117
JGUQDUKBUKFFRO-GGWOSOGESA-N
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
5356010
C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 239.0°C to 242.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3C(=NOH)C(=NOH)CH3
Quantity 100 g
Beilstein 01,772
Merck Index 15,327
Solubility Information Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene
Sublimation Point >70 °C
IUPAC Name N-[(2Z)-3-nitrosobut-2-en-2-yl]hydroxylamine
Formula Weight 116.12
Percent Purity 99+%
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethylglyoxime
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /