missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIMT1L Monoclonal Antibody (OTI2E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810140
This item is not returnable.
View return policy
Description
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3′-end of 18S rRNA in the 40S particle.Specifications
| DIMT1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UNQ2 | |
| DIMT1 | |
| Human recombit protein fragment corresponding to amino acids 1-238 of human DIMT1L produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2E6 | |
| Unconjugated | |
| DIMT1 | |
| DIM1, DIMT1L, HSA9761, HUSSY5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27292 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction